Orders LinkContactUsLinkComponent
Reference electrodes, ScienceLine
Reference electrodes, ScienceLine
Supplier:   SI Analytics
Reference electrodes, ScienceLine
Supplier:  SI Analytics
Certificates
Reference electrodes, ScienceLine
Reference electrodes, ScienceLine
5 Options Available Below
Product Details & Documents
Reference electrodes with glass shaft.
Product Family Options
Product Information
  • Type
    pH
    Temp.
    Material
    Electrolyte
    Diaphr.
    Ø×L
    Availability
    Price
  • SCOI1070046
    Single reference electrode, ScienceLine, Ag/AgCl reference system
    Single reference electrode, ScienceLine, Ag/AgCl reference system
    B 2920+ Ag/AgCl
    0 - 14
    −5...+100 °C
    Glass
    KCl 3 mol/l
    Platinum
    12×120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 193,00 €
    193,00 €
    1 * 1 ST
       193,00 €
  • SCOI1070044
    Single reference electrode, ScienceLine, Ag/AgCl reference system
    Single reference electrode, ScienceLine, Ag/AgCl reference system
    B 2820+ Ag/AgCl
    0 - 14
    −5...+100 °C
    Glass
    KCl 3 mol/l
    Ceramic
    12×120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 165,00 €
    165,00 €
    1 * 1 ST
       165,00 €
  • SCOI1070073
    Single reference electrode, ScienceLine, with standard taper NS 14,5, Ag/AgCl reference system
    Single reference electrode, ScienceLine, with standard taper NS 14,5, Ag/AgCl reference system
    B 3520+ Ag/AgCl
    0 - 14
    −5...+100 °C
    Glass
    KCl 3 mol/l
    Platinum
    10×103 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 229,00 €
    229,00 €
    1 * 1 ST
       229,00 €
  • SCOI1070070
    Single reference electrode, ScienceLine, with standard taper NS 14,5, Ag/AgCl reference system
    Single reference electrode, ScienceLine, with standard taper NS 14,5, Ag/AgCl reference system
    B 3420+ Ag/AgCl
    0 - 14
    −5...+100 °C
    Glass
    KCl 3 mol/l
    Ceramic
    10×103 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 229,00 €
    229,00 €
    1 * 1 ST
       229,00 €
  • SCOI1070075
    Single reference electrode, ScienceLine, with double electrolyte system, standard taper NS 14,5, Ag/AgCl reference system
    Single reference electrode, ScienceLine, with double electrolyte system, standard taper NS 14,5, Ag/AgCl reference system
    B 3920+ Ag/AgCl
    0 - 14
    −5...+100 °C
    Glass
    KCl 3 mol/l
    Ground joint
    10×103 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 415,00 €
    415,00 €
    1 * 1 ST
       415,00 €