ContactUsLinkComponent
Software for calorimeters, C 6040 CalWin
Software for calorimeters, C 6040 CalWin
Catalog # IKAA4040500
Supplier:  IKA
undefined
Software for calorimeters, C 6040 CalWin
Catalog # IKAA4040500
Supplier:  IKA
Specifications
  • Description:
    CalWin control and evaluation software, C 6040, for IKA® calorimeters
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
PC control and evaluation software for all IKA® calorimeters (C 200, C 2000, C 5000, C 7000, C 4000).
  • Print and save measurement protocols
  • Identify and record samples
  • Library functions
  • Data transfer through COM to EXCEL and ACCESS
  • Preprocessed work sheets for EXCEL (configurable by the user)
It requires a PC running any of the following operating systems: Windows 95/98/ME/NT/2000 or XP, and at least one free serial interface and 50 MB of available disk space.
Delivery: Supplied with a C 5041.10 connecting cable.