ContactUsLinkComponent
Table clamp
Catalog # USBE2051
undefined
Table clamp
Catalog # USBE2051
Specifications
  • For:
    Rods and square Rods
  • Spanning width:
    26 mm
  • Material:
    Aluminium alloy
  • Pk:
    1
Specifications
About this item
Large table clamp made of powder-coated aluminium alloy.
  • For rods and square rods up to 26 mm Ø
  • For benches up to 55 mm