Order Entry
Austria
ContactUsLinkComponent
 
 
3,5-Dinitrosalicylic acid 98%
 
undefined
 
3,5-Dinitrosalicylic acid 98%
 

 

  • Appearance:
    Yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    170 to 174 °C
  • Titration with NaOH:
    ≥97.5 %

 

 

Frequently Bought Together