Order Entry
Austria
ContactUsLinkComponent
4-Aminobenzoic acid
4-Aminobenzoic acid
Catalog # PROOMM0094.03
CAS Number:  
4-Aminobenzoic acid
Catalog # PROOMM0094.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :137.14 g/mol
Merck Index:13,00422
Boiling Pt:340 °C (1013 hPa)
Melting Pt:188 °C
Density:1.45 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:205-753-0
CAS Number:150-13-0
Formula:C₇H₇NO₂
MDL Number:MFCD00007894