
4-Aminobenzoic acid ≥99%, white powder cell culture reagent
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:150-13-0
:
Formula:C₇H₇NO₂
MDL Number:MFCD00007894
:137.14 g/mol
Merck Index:13,00422
Boiling Pt:340 °C (1013 hPa)
Melting Pt:180…190 °C
Density:1.374 g/mL (25 °C) (lit.)
EINECS:205-753-0
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥99%
- FTIR:Conforms to Standard
- UV/Vis: I max (water):275 ± 3 nm
- Loss on Drying:≤1.0%
Specification Test Results
Frequently Bought Together