Ammonium benzoate
Catalog # MOLEM62761848
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
Merck Index:12,00521
Melting Pt:197.8 °C
Density:1.26 g/cm³ (20 °C)
:139.15 g/mol
EINECS:217-468-9
:GHS07
Storage Temperature:Ambient
CAS Number:1863-63-4
Formula:C₇H₉NO₂
MDL Number:MFCD00013069