Orders LinkContactUsLinkComponent
Ostwald viscometers
Supplier:   SI Analytics
Ostwald viscometers
Supplier:  SI Analytics
Certificates
Ostwald viscometers
Ostwald viscometers
5 Options Available Below
Product Details & Documents
Ostwald viscometers suitable for measurements of transparent liquids manual measurements, foaming liquids and liquid mixture with highly volatile components.
Product Family Options
Product Information
  • Range
    Constant
    Length
    Ø int.
    Volume
    Availability
    Price
  • >0,3 mm²/s
    0,004 K
    220 mm
    0,30 mm
    3 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
  • >5,5 mm²/s
    0,07 K
    220 mm
    0,60 mm
    3 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
  • >10 mm²/s
    0,1 K
    220 mm
    0,70 mm
    3 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
  • >1 mm²/s
    0,01 K
    220 mm
    0,40 mm
    3 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
  • >2,5 mm²/s
    0,03 K
    220 mm
    0,50 mm
    3 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €