Orders LinkContactUsLinkComponent
Multi-channel pipettes, Multipet
Multi-channel pipettes, Multipet
Supplier:   KARTELL
Multi-channel pipettes, Multipet
Supplier:  KARTELL
Certificates
Multi-channel pipettes, Multipet
Multi-channel pipettes, Multipet
4 Options Available Below
Product Details & Documents
These multi-channel micropipettes with air cushion are ideal for dispensing in multiwell plates for ELISA dosages, molecular biology and DNA amplifications.
  • Volume can be easily selected by turning the emptying button or the cylinder
  • Large display ensures easy reading of the set volume value
  • Cones are indipendent, so the required force for securing the 8 or 12 tips is uniformly distributed
  • Minimum pressure required for inserting and ejecting tips
  • Movement of the cones allows the micropipette to oscillate during the loading operations
  • Lower part can freely turn 360°, the ejector is easily adjustable to house a large variety of tips and can be removed for cleaning
  • Fully autoclavable at +121 °C for 20 minutes, without having to recalibrate them
Certifications: EC/IVD-compliant, in accordance with EN IS 8655
Delivery information: Supplied with individual calibration certificates.
Product Family Options
8-Channel
Product Information
  • Volume
    Accuracy
    Imprecision
    Availability
    Price
  • 50 - 300 µl
    ±1,6 (50 µl); ±1,0 (300 µl) %
    ≤ ±3,0 (50 µl); ±1,0 (300 µl) %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 370,00 €
    1 370,00 €
    1 * 1 ST
       1 370,00 €
  • 5 - 50 µl
    ±4,0 (5 µl); ±1,6 (50 µl) %
    ±1,5 (5 µl); ±0,8 (50 µl) %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 370,00 €
    1 370,00 €
    1 * 1 ST
       1 370,00 €
12-Channel
Product Information
  • Volume
    Accuracy
    Imprecision
    Availability
    Price
  • 50 - 300 µl
    ±1,6 (50 µl); ±1,0 (300 µl) %
    ≤ ±3,0 (50 µl); ±1,0 (300 µl) %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 740,00 €
    1 740,00 €
    1 * 1 ST
       1 740,00 €
  • 5 - 50 µl
    ±4,0 (5 µl); ±1,6 (50 µl) %
    ±1,5 (5 µl); ±0,8 (50 µl) %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 780,00 €
    1 780,00 €
    1 * 1 ST
       1 780,00 €
Accessories
Product Information
  • Catalog #
    Colour
    Availability
    Price
  • Stand for 3 pipettors, 145×70 mm
    Stand for 3 pipettors, 145×70 mm
    KART1315
    Transparent
    Log in for availability
    1 * 1 ST - 81,90 €