Order Entry
Austria
Orders LinkContactUsLinkComponent
Vanillin ≥99.5%

Vanillin ≥99.5%

Glass bottle for solids, 1 kg
Artikelnr. MOLE61624662-1KG
Irritant
Marketsource
Anbieter:   Molekula
Vanillin ≥99.5%
Artikelnr. MOLE61624662-1KG
Anbieter:  Molekula
Marketsource
CAS Nummer:  121-33-5
Vanillin ≥99.5%
Vanillin ≥99.5%

Glass bottle for solids, 1 kg

CAS Nummer:  121-33-5
Preis
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:12,10068
  • Boiling Pt:353 °C (1013 hPa)
  • Melting Pt:81…83 °C
  • Storage Temperature:Ambient
  • EINECS:204-466-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Cat. No.:
    MOLE61624662-1KG

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.