Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pipettes
Pipette Bulbs
Pipette bulb
Pipette bulb
Pipette bulb
Catalog # KART208
Supplier:
KARTELL
Expand image
Pipette bulb
Catalog # KART208
Supplier:
KARTELL
Specifications
Colour:
Red
Pk:
1
Specifications
About this item
Product Details
Pipette bulb made of red rubber and silicone.
Product Details