ContactUsLinkComponent
Pipette bulb, with adapter
Pipette bulb, with adapter
Catalog # KART200
Supplier:  KARTELL
undefined
Pipette bulb, with adapter
Catalog # KART200
Supplier:  KARTELL
Specifications
  • Colour:
    Red
  • Pk:
    1
Specifications
About this item
Moulded pipette bulb made of synthetic rubber.
  • Three glass ball valves
  • Can be controlled by finger pressure
  • No metal parts to corrode
  • Can be used with all liquids except those that attack rubber
  • Use on bulb or cylindrical pipettes of different capacities
Delivery: Supplied with adapter for pipettes with capacities more than 20 ml.