Orders LinkContactUsLinkComponent
Sample containers with screw cap
Sample containers with screw cap
Supplier:   KARTELL
Sample containers with screw cap
Supplier:  KARTELL
Certificates
Sample containers with screw cap
Sample containers with screw cap
4 Options Available Below
Product Details & Documents
PFA, translucent, with PFA screw cap.
  • Very good chemical resistance
  • Very good temperature resistance
For sample collection, transport and storage.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Availability
    Price
  • 30 ml
    38 mm
    54 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 127,00 €
    127,00 €
    1 * 1 ST
       127,00 €
  • 60 ml
    38 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 153,00 €
    153,00 €
    1 * 1 ST
       153,00 €
  • 90 ml
    54 mm
    62 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 168,00 €
    168,00 €
    1 * 1 ST
       168,00 €
  • 180 ml
    54 mm
    112 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 194,00 €
    194,00 €
    1 * 1 ST
       194,00 €