ContactUsLinkComponent
Sedimentation cones, Imhoff type, DURAN®
Sedimentation cones, Imhoff type, DURAN®
Catalog # SCOT214015403
Supplier:  DWK Life Sciences
undefined
Sedimentation cones, Imhoff type, DURAN®
Catalog # SCOT214015403
Supplier:  DWK Life Sciences
Specifications
  • Type:
    Without stopcock
  • Description:
    Imhoff cones, scale up to 1000 ml, ring mark at 1000 ml
  • Pk:
    1
Specifications
About this item
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • Very good chemical resistance
  • Without stopcock
The conical sedimentation cone with graduation according to Imhoff allows a quantification of sediments from a suspension (e.g. in waste water).
Certifications: According to DIN 12672.