Extraction thimbles
Supplier: Lenz Laborglas
Jump to:
About this item
DURAN®, Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Glass insert with bores for solids.
3 Options Available Below
Product Details & Documents
DURAN®, Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Glass insert with bores for solids.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Product Family Options
Product Information
- AvailabilityPrice
- LENZ05431201Extraction insert for solids, for extractor 70 ml capacity (100 ml volume)Extraction insert for solids, for extractor 70 ml capacity (100 ml volume)Log in for availability1 * 1 ST38,60 €
- LENZ05431202Extraction insert for solids, for extractor 100 ml capacity (250 ml volume)Extraction insert for solids, for extractor 100 ml capacity (250 ml volume)Log in for availability1 * 1 ST41,60 €
- LENZ05431203Extraction insert for solids, for extractor 250 ml capacity (500 ml volume)Extraction insert for solids, for extractor 250 ml capacity (500 ml volume)Log in for availability1 * 1 ST49,30 €