Nalgene® Diagnostics Bottles, Narrow Neck, Round, HDPE
Bottles without closure, used to store and ship small volumes of valuable reagents.
- Nonsterile
- Durable, uniform walls are resistant to splitting or puncturing
- Leakproof when assembled with Nalgene closure
Natural, translucent high-density polyethylene, durable, uniform walls resistant to splitting or puncturing, A sample product certificate providing cytotoxicity, USP Class VI, FDA compliance 21 CFR and other regulatory compliance information is available or can be requested from our certificates of compliance database.
: Manufactured in compliance with ISO 13485:2016 quality requirements. Diagnostic bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP class VI.
: Coloured closures are ordered seperately.
- Catalog No:
- NALG362002-9025
- NALG362002-9050
- NALG362002-9125
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Frequently Bought Together