Nalgene® Diagnostics Bottles, Narrow Neck, Round, PP
Supplier: Thermo Fisher Scientific
Certificates
About this item
Bottles with polypropylene closure used for safely storage and ship small volume samples and valuable reagents.
- Nonsterile
- Made up of natural translucent PPCO
- Leakproof
Thermo Scientific™ Nalgene™ Natural PPCO diagnostic bottles with closures are used in test kits and many small-volume applications. These autoclavable and durable polypropylene copolymer bottles are tray packed with closure for easy handling and feature uniform walls resistant to splitting or puncturing.
Certifications: Compliance with ISO 13485:2003 and 21CFR177.1520, USP class VI.
Packaging: Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton.
Specifications
- Catalog No:
- NALG322006-9050
- NALG322006-9025
- NALG322006-9125
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)