Orders LinkContactUsLinkComponent
Test tubes
Test tubes
Supplier:   KARTELL
Test tubes
Supplier:  KARTELL
Certificates
Test tubes
2 Options Available Below
Product Details & Documents
PTFE, white.
  • Excellent chemical resistance
  • Very good mechanical resistance
  • Very good temperature resistance
  • Suitable for foodstuff
Will withstand temperatures continuously up to 250 °C and for short intervals up to 300 °C.
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Ø int.
    Availability
    Price
  • 10 ml
    120 mm
    18 mm
    15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,80 €
    58,80 €
    1 * 1 ST
       58,80 €
  • 15 ml
    100 mm
    18 mm
    15 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,10 €
    55,10 €
    1 * 1 ST
       55,10 €