Order Entry
Austria
ContactUsLinkComponent
 
Cholecalciferol (Vitamin D3)
Catalog # 47763.
Supplier:  Merck
CAS Number:  
undefined
Cholecalciferol (Vitamin D3)
Catalog # 47763.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Pk Info:
    100 mg

Specifications

About this item

 :
 :384.65 g/mol
EINECS:200-673-2
UN:2811
ADR:6.1,II
CAS Number:67-97-0
Formula:C₂₇H₄₄O
MDL Number:MFCD00078131