Cholecalciferol (Vitamin D3), Sigma-Aldrich®
Catalog # SIALC9756-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:384.65 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:200-673-2
UN:2811
CAS Number:67-97-0
Formula:C₂₇H₄₄O
MDL Number:MFCD00078131