Calcium citrate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5785-44-4
:
Formula:C₁₂H₁₀Ca₃O₁₄·4H₂O
MDL Number:MFCD00150786
:570.5 g/mol
:M-Clarity™ quality level = MQ200
EINECS:212-391-7
Frequently Bought Together