3-Methyl-2-cyclohexen-1-ol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 3-Methylcyclohex-2-enol, 3-Methylcyclohex-2-en-1-ol, 3-Methylcyclohex-2-ene-1-ol
CAS Number:21378-21-2
:
Formula:C₇H₁₂O
MDL Number:MFCD00001553
Molecular Weight:112.17 g/mol
:M-Clarity™ quality level = MQ100
EINECS:244-353-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together