
Zinc(II) acetylacetonate, Sigma-Aldrich®
Catalog # 8.08803.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:6.9 g/l (20 °C)
- Melting Point:124 - 126 °C
- Molar Mass:263.61 g/mol
- Bulk Density:500 kg/m3
- Boiling Point:129 - 131 °C (13 hPa)
- Vapor Pressure:1 hPa (20 °C)
- pH:10 (4 g/l, H2O, 20 °C)
- Ignition Point:580 °C
Specification Test Results
About this item
:
:263.61 g/mol
:M-Clarity™ quality level = MQ200
EINECS:237-860-3
:GHS07
Storage Temperature:Ambient
CAS Number:14024-63-6
Formula:C₁₀H₁₄O₄Zn
MDL Number:MFCD00000035