Trifluoroacetic acid 25% (v/v) in aqueous solution, Supelco®
Catalog # 1.08218.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.12 g/cm3 (20 °C)
- pH:(H2O, 20 °C) strongly acid
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
About this item
Synonyms: TFA
:
:114.02 g/mol
ADR:8,I
:M-Clarity™ quality level = MQ200
EINECS:200-929-3
UN:2699
:GHS05
Storage Temperature:Ambient
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169