
D-Cycloserine, off-white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:68-41-7
:
Formula:C₃H₆N₂O₂
MDL Number:MFCD00005353
:102.09 g/mol
EINECS:200-688-4
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- Potency:≥900 µg/mg
- FTIR:Conforms to Standard
- pH (2% aq soln):5,5 - 6,5
- Solubility (5% aq soln):Clear, Colorless to Slightly Pink to Yellow Solution
- Typical Working Concentration:100-200 µg/ml. Prepare just before use using 0,1 M sodium phosphate buffer, pH 8,0,
Specification Test Results
Frequently Bought Together