Organic reference standard, Chloroethylene 1000 µg/ml in methanol, Dr Ehrenstorfer
EHERY17923000ME
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:75-01-4
- Description:Chloroethylene 1,000 µg/ml in methanol
- Formula:C₂H₃Cl
- Description:Chloroethylene