Organic reference standard, 1,2,4-Trimethylbenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17880800ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:95-63-6
- Description:1,2,4-Trimethylbenzene 100 µg/ml in methanol
- Formula:C₉H₁₂
- Description:1,2,4-Trimethylbenzene