Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 1,1,2-Trichlorotrifluoroethane 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, 1,1,2-Trichlorotrifluoroethane 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA17788300ME
 :  
Organic reference standard, 1,1,2-Trichlorotrifluoroethane 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA17788300ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    76-13-1
  • Description:
    1,1,2-Trichlorotrifluoroethane 100 µg/ml in methanol
  • Formula:
    C₂Cl₃F₃
  • Description:
    1,1,2-Trichlorotrifluoroethane