Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Bromoform 100 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, Bromoform 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA17665500ME
 :  
Organic reference standard, Bromoform 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA17665500ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    75-25-2
  • Description:
    Bromoform 100 µg/ml in methanol
  • Formula:
    CHBr₃
  • Description:
    Bromoform