Organic reference standard, 1-Triacontanol 100 µg/ml in tert-butyl methyl ether, Dr Ehrenstorfer
Catalog # EHERXA17609200MB
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:593-50-0
- Description:1-Triacontanol 100 µg/ml in tert-butyl methyl ether
- Formula:C₃₀H₆₂O
- Description:1-Triacontanol