Organic reference standard, Norflurano 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17404000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:811-97-2
- Description:Norflurano 100 µg/ml in methanol
- Formula:C₂H₂F₄
- Description:Norflurano