Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 1,2-Dichloropropane 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, 1,2-Dichloropropane 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12480000CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1,2-Dichloropropane 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12480000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    78-87-5
  • Description:
    1,2-Dichloropropane 100 µg/ml in cyclohexane
  • Formula:
    C₃H₆Cl₂
  • Description:
    1,2-Dichloropropane

Specifications

About this item