Organic reference standard, Chlorodifluoromethane 100 µg/ml in methanol, Dr Ehrenstorfer
EHERXA11404400ME
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:75-45-6
- Description:Chlorodifluoromethane 100 µg/ml in methanol
- Formula:CHClF₂
- Description:Chlorodifluoromethane