Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 1-Methylpyrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, 1-Methylpyrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20901000CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1-Methylpyrene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL20901000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    2381-21-7
  • Description:
    1-Methylpyrene 10 µg/ml in cyclohexane
  • Formula:
    C₁₇H₁₂
  • Description:
    1-Methylpyrene

Specifications

About this item