Organic reference standard, Fluoranthene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20795000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:206-44-0
- Description:Fluoranthene 10 µg/ml in acetonitrile
- Formula:C₁₆H₁₀
- Description:Fluoranthene