Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 1,2,4,5-Tetramethylbenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Organic reference standard, 1,2,4,5-Tetramethylbenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17412500CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 1,2,4,5-Tetramethylbenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17412500CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    95-93-2
  • Description:
    1,2,4,5-Tetramethylbenzene 10 µg/ml in cyclohexane
  • Formula:
    C₁₀H₁₄
  • Description:
    1,2,4,5-Tetramethylbenzene

Specifications

About this item