Organic reference standard, α-Linolenic acid 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL14635600ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:463-40-1
- Description:α-Linolenic acid 10 µg/ml in methanol
- Formula:C₁₈H₃₀O₂
- Description:α-Linolenic acid