Organic reference standard, Oxybenzone 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Artikelnr. EHERL14232500CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:131-57-7
- Description:Oxybenzone 10 µg/ml in cyclohexane
- Formula:C₁₄H₁₂O₃
- Description:Oxybenzone