
Methyl-3-bromo-4-methoxybenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35450-37-4
:
Formula:C₉H₉BrO₃
MDL Number:MFCD00210463
:245.07 g/mol
Melting Pt:95…98 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together