(4-Hydroxyphenyl)acetic acid methyl ester 99%
:14199-15-6
:
:C₉H₁₀O₃
:MFCD00002387
:166.18 g/mol
:162…163 °C (5 mmHg)
:55…58 °C
:238-050-2
:GHS07
- Appearance:White to light yellow Crystals
- Infrared spectrum:Conforms
- Melting point:55 to 58 °C
- GC:≥98.5 %
Frequently Bought Together