ContactUsLinkComponent
Screw caps for narrow neck bottles, square, with thread, for bottles "SafeGrip" series
Screw caps for narrow neck bottles, square, with thread, for bottles "SafeGrip" series
Catalog # KAUT805-85941
Supplier:  Kautex
undefined
Screw caps for narrow neck bottles, square, with thread, for bottles "SafeGrip" series
Catalog # KAUT805-85941
Supplier:  Kautex
Specifications
  • Thread:
    32 mm
  • Colour:
    Blue
  • Description:
    Screw closure, with PTFE insert and tamper evident ring
  • Material:
    PP
  • Pk:
    1
  • For bottles:
    50 - 500 ml