Order Entry
Austria
ContactUsLinkComponent
 
 
 
Ethanol absolute, EMPARTA® ACS analytical reagent, Supelco®
 :  Merck
    
 
undefined
 
 
Ethanol absolute, EMPARTA® ACS analytical reagent, Supelco®
 :  Merck
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

 

 

  • Solubility:
    (20 °C) completely miscible
  • Melting Point:
    -114.5 °C
  • Molar Mass:
    46.07 g/mol
  • Boiling Point:
    78.3 °C (1013 hPa)
  • Vapor Pressure:
    59 hPa (20 °C)
  • Flash Point:
    12 °C
  • Refractive Index:
    1.36
  • Explosion Limit:
    3.5 - 15 %(V)
  • Density:
    0.790 - 0.793 g/cm3 (20 °C)
  • pH:
    7.0 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    425 °C

 

Frequently Bought Together