Orders LinkContactUsLinkComponent
Conical separating funnels, DURAN®
Conical separating funnels, DURAN®
Supplier:   DWK Life Sciences
Conical separating funnels, DURAN®
Supplier:  DWK Life Sciences
Certificates
Conical separating funnels, DURAN®
Conical separating funnels, DURAN®
10 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with NS-PE stopper.
  • Very good chemical resistance
  • Very good temperature resistance
  • Wwithout graduation complies to DIN ISO 4800
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance.
Product Family Options
With solid glass key, without graduation
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 50 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,60 €
    77,60 €
    1 * 1 ST
       77,60 €
  • 100 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,40 €
    80,40 €
    1 * 1 ST
       80,40 €
  • 250 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,10 €
    90,10 €
    1 * 1 ST
       90,10 €
  • 500 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • 1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
  • 2000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 201,00 €
    201,00 €
    1 * 1 ST
       201,00 €
With PTFE key and graduation
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 500 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €
  • 1000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 152,00 €
    152,00 €
    1 * 1 ST
       152,00 €
  • 100 ml
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,40 €
    80,40 €
    1 * 1 ST
       80,40 €
  • 250 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,10 €
    90,10 €
    1 * 1 ST
       90,10 €