Bioassay dishes, 4-well, for in vitro fertilisation, Nunc™
PS, sterile, flat bottom, with lid. These fully certified dishes are specifically intended for in vitro fertilisation (IVF) use.
- Produced under ISO 13485 with full lot traceability
- Certificates of conformity based on a 1-cell stage mouse embryo toxicity test, confirm sterility (SAL 10⁶), non pyrogenicity (LAL test), and that the material has passed USP class VI and MEA test requirements
Frequently Bought Together