Gas distribution tubes with lateral ring
Supplier: LENZ LABORGLAS GMBH & CO.KG
Certificates
About this item
Gas distribution tubes with lateral ring made of borosilicate glass 3,3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion
Frequently Bought Together