Orders LinkContactUsLinkComponent
Filter funnels
Filter funnels
Supplier:   Lenz Laborglas
Filter funnels
Supplier:  Lenz Laborglas
Certificates
Filter funnels
24 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Documents
Product Family Options
Product Information
  • Capacity
    Plate Ø
    Porosity
    Availability
    Price
  • 50 ml
    35 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €
  • 50 ml
    35 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €
  • 50 ml
    35 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €
  • 50 ml
    35 mm
    4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,00 €
    32,00 €
    1 * 1 ST
       32,00 €
  • 75 ml
    45 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,60 €
    33,60 €
    1 * 1 ST
       33,60 €
  • 75 ml
    45 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,60 €
    33,60 €
    1 * 1 ST
       33,60 €
  • 75 ml
    45 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,60 €
    33,60 €
    1 * 1 ST
       33,60 €
  • 75 ml
    45 mm
    4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,50 €
    37,50 €
    1 * 1 ST
       37,50 €
  • 125 ml
    60 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,80 €
    45,80 €
    1 * 1 ST
       45,80 €
  • 125 ml
    60 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,80 €
    45,80 €
    1 * 1 ST
       45,80 €
  • 125 ml
    60 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,80 €
    45,80 €
    1 * 1 ST
       45,80 €
  • 125 ml
    60 mm
    4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,40 €
    52,40 €
    1 * 1 ST
       52,40 €
  • 500 ml
    95 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,20 €
    83,20 €
    1 * 1 ST
       83,20 €
  • 500 ml
    95 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,20 €
    83,20 €
    1 * 1 ST
       83,20 €
  • 500 ml
    95 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,20 €
    83,20 €
    1 * 1 ST
       83,20 €
  • 500 ml
    95 mm
    4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 90,30 €
    90,30 €
    1 * 1 ST
       90,30 €
  • 1000 ml
    120 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
  • 1000 ml
    120 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
  • 1000 ml
    120 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 145,00 €
    145,00 €
    1 * 1 ST
       145,00 €
  • 1000 ml
    120 mm
    4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 187,00 €
    187,00 €
    1 * 1 ST
       187,00 €
  • 4000 ml
    179 mm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 485,00 €
    485,00 €
    1 * 1 ST
       485,00 €
  • 4000 ml
    179 mm
    2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 485,00 €
    485,00 €
    1 * 1 ST
       485,00 €
  • 4000 ml
    179 mm
    3
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 485,00 €
    485,00 €
    1 * 1 ST
       485,00 €
  • 4000 ml
    179 mm
    4
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 563,00 €
    563,00 €
    1 * 1 ST
       563,00 €