Kolben, flacher Boden, enghalsig, mit Bördelrand
DURAN®, borosilicate glass 3,3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area. Flasks with neck diameter of 65 mm have a reinforced rims.
: Acc. to DIN ISO 1773 and in addition to standard.
Frequently Bought Together