Beakers, tall form
Catalog # LENZ07004070
Supplier: LENZ LABORGLAS GMBH & CO.KG
Supplier #: 07004070
Log in for availability
About this item
DURAN® borosilicate glass, with spout.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area.
Frequently Bought Together