Tongs, Chemware®
Catalog # VERND1069290
Supplier: Saint Gobain Life Sciences
Specifications
- Length:240 mm
- Pk:1
Specifications
About this item
These stainless steel tongs are covered with an extra-heavy, non stick PTFE coating. For handling hot, cold, corrosive or sticky materials.
- No loss of sample through sticking
- Will not scratch or damage delicate surfaces
- Contoured jaw enables handling of crucibles and other objects