5-Bromovanillin ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 3-Bromo-4-hydroxy-5-methoxybenzaldehyde
CAS Number:2973-76-4
:
Formula:C₈H₇BrO₃
MDL Number:MFCD00006940
Boiling Pt:314
Molecular Weight:231.05 g/mol
Melting Pt:162…166 °C
EINECS:221-016-6
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together